Tisk…

4,4'-Dimethylbenzophenone ≥98%

Dodavatel: Thermo Fisher Scientific

A14111.06 A14111.14 A14111.22
A14111.06EA 1150 CZK
A14111.06 A14111.14 A14111.22
4,4'-Dimethylbenzophenone ≥98%
4,4'-Dimethylbenzophenone

Purity: 98+% CAS No.: 611-97-2 Molecular Formula: C15H14O Formula Weight: 210.28 Melting Point: 92-96° Boiling Point: 200°/17mm MDL No.: MFCD00017214

Vzorec: C₁₅H₁₄O
Bod varu: 200 °C (17 mmHg)
Bod tání: 92…96 °C
Hustota: 1,18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00017214
CAS číslo: 611-97-2
EINECS: 210-287-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR