Tisk…

4-Cyanobenzenesulphonyl chloride ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A14081.14 A14081.03 A14081.06
A14081.14EA 21490 CZK
A14081.14 A14081.03 A14081.06
4-Cyanobenzenesulphonyl chloride ≥97%
4-Cyanobenzenesulphonyl chloride

Purity: 97% CAS No.: 49584-26-1 Molecular Formula: C7H4ClNO2S Formula Weight: 201.63 Melting Point: 107-110° Beilstein Reference: 777939 UN No.: 3261 MDL No.: MFCD00052337

Vzorec: C₇H₄ClNO₂S
Bod tání: 107…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00052337
CAS číslo: 49584-26-1
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR