Tisk…

4-tert-Butylbenzoic acid ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 4-(tert-Butyl)benzoic acid

A13997.30 A13997.0B A13997.0I
A13997.30EA 841 CZK
A13997.30 A13997.0B A13997.0I
4-tert-Butylbenzoic acid ≥99%
4-tert-Butylbenzoic acid

Purity: 99+% CAS No.: 98-73-7 Molecular Formula: C11H14O2 Formula Weight: 178.23 Flash Point: 180°(356°F) Melting Point: 164-168° Boiling Point: 280° dec. Beilstein Reference: 607545 UN No.: 2811 MDL No.: MFCD00002563

Vzorec: C₁₁H₁₄O₂
MW: 178.23
Bod varu: 280 °C (rozkl.)
Bod tání: 164…168 °C
Hustota: 0.6
Bod vzplanutí: 180 °C (356 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002563
CAS číslo: 98-73-7
EINECS: 202-696-3
UN: 2811
ADR: 6.1 III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR