Tisk…

aceton-dimethylacetal ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A13810.0F A13810.AE A13810.AP A13810.PI
A13810.0FEA 7110 CZK
A13810.0F A13810.AE A13810.AP A13810.PI
aceton-dimethylacetal ≥98%
aceton-dimethylacetal

Purity: 98% CAS No.: 77-76-9 Molecular Formula: (CH3O)2C(CH3)2 Formula Weight: 104.15 Flash Point: -11°(12°F) Physical Form: Liquid Density: 0.848 Refractive Index: 1.3780 Appearance: Colourless Odour: Sweet Melting Point: -47° Boiling Point: 79-81° Beilstein Reference: 635678 UN No.: 3271 MDL No.: MFCD00008479

Vzorec: C₅H₁₂O₂
Bod varu: 79…81 °C
Bod tání: –47 °C
Hustota: 0,848
Bod vzplanutí: –11 °C (–12 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008479
CAS číslo: 77-76-9
EINECS: 201-056-0
UN: 3271
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR