Tisk…

3,5-Dibromosalicylaldehyde ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A13729.09 A13729.18
A13729.09EA 2300 CZK
A13729.09 A13729.18
3,5-Dibromosalicylaldehyde ≥98%
3,5-Dibromosalicylaldehyde

Purity: 98% CAS No.: 90-59-5 Molecular Formula: C7H4Br2O2 Formula Weight: 279.93 Melting Point: 81-84° Beilstein Reference: 1424739 Merck Reference: 14,3027 MDL No.: MFCD00003318

Vzorec: C₇H₄Br₂O₂
Bod varu: 261 °C (1013 hPa)
Bod tání: 81…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00003318
CAS číslo: 90-59-5
EINECS: 202-003-4
Merck Index: 14,3027

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR