Tisk…

4-Piperidone monohydrate hydrochloride 98%

Dodavatel: Thermo Fisher Scientific

Synonyms: 4,4-Piperidinediol Hydrochloride, 4,4-Dihydroxypiperidine Hydrochloride, 4-Ketopiperidine Monohydrate Hydrochloride, 4-Piperidinone monohydrate hydrochloride, Piperidin-4-one hydrate hydrochloride, 4-Azacyclohexanone Monohydrate Hydrochloride

A13726.09 A13726.18
A13726.09EA 1360 CZK
A13726.09 A13726.18
4-Piperidone monohydrate hydrochloride 98%
4-Piperidone monohydrate hydrochloride

Purity: 98% CAS No.: 40064-34-4 Molecular Formula: C5H9NO.HCl.H2O Formula Weight: 153.61 (135.59anhy) Melting Point: 94-100° Beilstein Reference: 3916558 MDL No.: MFCD00012776

Vzorec: C₅H₉NO
MW: 99.13 g/mol (free base)
Bod tání: 94…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00012776
CAS číslo: 40064-34-4
EINECS: 254-779-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR