Tisk…

1-Butylboronic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Butylboronic acid, n-Butylboronic acid

A13725.03 A13725.06 A13725.14
A13725.03EA 951 CZK
A13725.03 A13725.06 A13725.14
1-Butylboronic acid ≥98%
1-Butylboronic acid

Purity: 98% CAS No.: 4426-47-5 Molecular Formula: CH3CH2CH2CH2B(OH)2 Formula Weight: 101.94 Storage Temperature: 2°C to 8°C Melting Point: 92-96° Beilstein Reference: 1733489 MDL No.: MFCD00002106

Vzorec: C₄H₁₁BO₂
MW: 101.94
Bod tání: 92…96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002106
CAS číslo: 4426-47-5
EINECS: 224-607-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR