Tisk…

myo-Inozitol ≥98%

Dodavatel: Thermo Fisher Scientific

A13586.0E A13586.22 A13586.36
A13586.0EEA 18120 CZK
A13586.0E A13586.22 A13586.36
myo-Inozitol ≥98%
myo-Inozitol

Purity: 98+% CAS No.: 87-89-8 Molecular Formula: C6H12O6 Formula Weight: 180.16 Physical Form: Cryst powder Density: 1.75 Appearance: White Odour: Odourless Melting Point: 224-227° Beilstein Reference: 1907329 Merck Reference: 14,4978 MDL No.: MFCD00077932


A lipotropic agent

Vzorec: C₆H₁₂O₆
Bod varu: 291 °C (1013 hPa)
Bod tání: 224…227 °C
Hustota: 1,75
Storage Temperature: Ambient
MDL Number: MFCD00077932
CAS číslo: 87-89-8
EINECS: 201-781-2
Merck Index: 14,4978

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR