Tisk…

Piperine 98%

Dodavatel: Thermo Fisher Scientific
Varování

A13510.06 A13510.14 A13510.22
A13510.06EA 1200 CZK
A13510.06 A13510.14 A13510.22
Piperine 98%
Piperine

Purity: 98% CAS No.: 94-62-2 Molecular Formula: C17H19NO3 Formula Weight: 285.34 Melting Point: 128-133° Beilstein Reference: 90741 MDL No.: MFCD00005839

Vzorec: C₁₇H₁₉NO₃
Bod tání: 128…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00005839
CAS číslo: 94-62-2
EINECS: 202-348-0
Merck Index: 14,7472

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR