Tisk…

1-Cetylpyridiniumchlorid monohydrát ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A13499.14 A13499.22 A13499.36
A13499.14EA 793 CZK
A13499.14 A13499.22 A13499.36
1-Cetylpyridiniumchlorid monohydrát ≥98%
1-Cetylpyridiniumchlorid monohydrát

Purity: 98% CAS No.: 6004-24-6 Molecular Formula: C21H38ClN.H2O Formula Weight: 358.02 (339.99anhy) Melting Point: 83-86° Solubility: Soluble in water, alcohol, chloroform Beilstein Reference: 3578606 Merck Reference: 14,2032 UN No.: 2811 MDL No.: MFCD00149977

Vzorec: C₂₁H₃₈ClN·1H₂O
Bod tání: 83…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00149977
CAS číslo: 6004-24-6
EINECS: 204-593-9
UN: 2811
ADR: 6.1,II
Merck Index: 14,2032

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR