Tisk…

3-Nitrobenzoic acid ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A13494.0B A13494.0I A13494.30
A13494.0BEA 3330 CZK
A13494.0B A13494.0I A13494.30
3-Nitrobenzoic acid ≥99%
3-Nitrobenzoic acid

Purity: 99%
CAS No.: 121-92-6
Molecular Formula: C7H5NO4
Formula Weight: 167.12
Melting Point: 140-143°
Beilstein Reference: 908644
Merck Reference: 14,6589
MDL No.: MFCD00007251

Vzorec: C₇H₅NO₄
Bod varu: 341 °C (1013 hPa)
Bod tání: 140…143 °C
Hustota: 1,494
Storage Temperature: Ambient
MDL Number: MFCD00007251
CAS číslo: 121-92-6
EINECS: 204-508-5
Merck Index: 14,6589

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR