Tisk…

(±)-4-Fluoromandelic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A13391.03 A13391.06 A13391.14
A13391.03EA 563 CZK
A13391.03 A13391.06 A13391.14
(±)-4-Fluoromandelic acid ≥98%
(±)-4-Fluoromandelic acid

Purity: 98% CAS No.: 395-33-5 Molecular Formula: C8H7FO3 Formula Weight: 170.14 Melting Point: 133-138° Beilstein Reference: 2211001

Vzorec: C₈H₇FO₃
Bod tání: 133…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00044402
CAS číslo: 395-33-5
EINECS: 206-899-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR