Tisk…

(±)-4-Bromomandelic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A12978.06 A12978.14 A12978.22
A12978.06EA 1400 CZK
A12978.06 A12978.14 A12978.22
(±)-4-Bromomandelic acid ≥98%
(±)-4-Bromomandelic acid

Purity: 98+% CAS No.: 6940-50-7 Molecular Formula: C8H7BrO3 Formula Weight: 231.05 Melting Point: 115-119° Beilstein Reference: 1567909 Merck Reference: 14,1424 MDL No.: MFCD00004232

Vzorec: C₈H₇BrO₃
Bod varu: 344 °C (1013 hPa)
Bod tání: 115…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00004232
CAS číslo: 6940-50-7
EINECS: 230-085-1
Merck Index: 14,1424

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR