Tisk…

Octan zinečnatý dihydrát ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Acetic acid zinc salt dihydrate, Zinc (II) acetate dihydrate, Zinc diacetate dihydrate

A12909.0B A12909.0I A12909.22 A12909.36 A12909.A4
A12909.0BEA 2640 CZK
A12909.0B A12909.0I A12909.22 A12909.36 A12909.A4
Octan zinečnatý dihydrát ≥97%
Octan zinečnatý dihydrát

Purity: 97+% CAS No.: 5970-45-6 Molecular Formula: Zn(OOCCH3)2.2H2O Formula Weight: 219.50 (183.47anhy) Odour: Weak, characteristic pH: 6-7 Melting Point: 237° Beilstein Reference: 3732513 Merck Reference: 14,10128 UN No.: 3077 MDL No.: MFCD00066961

Vzorec: (H₃CCOO)₂Zn·2H₂O
MW: 219.51
Bod tání: 237 °C
Hustota: 1.718
Storage Temperature: Ambient
MDL Number: MFCD00066961
CAS číslo: 5970-45-6
EINECS: 209-170-2
UN: 3077
ADR: 9 III
Merck Index: 12,10256

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR