Tisk…

Methyl-3-aminothiophene-2-carboxylate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A12802.14 A12802.06 A12802.22
A12802.14EA 3920 CZK
A12802.14 A12802.06 A12802.22
Methyl-3-aminothiophene-2-carboxylate ≥98%
Methyl-3-aminothiophene-2-carboxylate

Purity: 98+% CAS No.: 22288-78-4 Molecular Formula: C6H7NO2S Formula Weight: 157.19 Melting Point: 63-67° Boiling Point: 100-102°/0.1mm MDL No.: MFCD00009765

Vzorec: C₆H₇NO₂S
Bod varu: 100…102 °C (0,1 mmHg)
Bod tání: 63…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00009765
CAS číslo: 22288-78-4
EINECS: 244-895-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR