Tisk…

Oxalacetic acid ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A12739.06 A12739.14 A12739.22
A12739.06EA 3010 CZK
A12739.06 A12739.14 A12739.22
Oxalacetic acid ≥97%
Oxalacetic acid

Purity: 97% CAS No.: 328-42-7 Molecular Formula: C4H4O5 Formula Weight: 132.07 Storage Temperature: -30°C to -10°C Flash Point: 88°(190°F) Melting Point: 161° dec. Beilstein Reference: 1705475 Merck Reference: 14,6909 UN No.: 3261 MDL No.: MFCD00002592

Vzorec: C₄H₄O₅
Bod varu: 310 °C (1013 hPa)
Bod vzplanutí: 88 °C (190 °F)
Storage Temperature: Freezer
MDL Number: MFCD00002592
CAS číslo: 328-42-7
EINECS: 206-329-8
UN: 3261
ADR: 8,II
Merck Index: 14,6909

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR