Tisk…

4-Hydroxybenzhydrazide ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A12702.14 A12702.22 A12702.36
A12702.14EA 1380 CZK
A12702.14 A12702.22 A12702.36
4-Hydroxybenzhydrazide ≥98%
4-Hydroxybenzhydrazide

Purity: 98% CAS No.: 5351-23-5 Molecular Formula: C7H8N2O2 Formula Weight: 152.15 Melting Point: ca 267° dec. Beilstein Reference: 473052

Vzorec: C₇H₈N₂O₂
Bod varu: 370 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00007605
CAS číslo: 5351-23-5
EINECS: 226-326-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR