Tisk…

Kyselina 2-thiobarbiturová ≥98%

Dodavatel: Thermo Fisher Scientific

A12681.14 A12681.22 A12681.36
A12681.14EA 1600 CZK
A12681.14 A12681.22 A12681.36
Kyselina 2-thiobarbiturová ≥98%
Kyselina 2-thiobarbiturová

Purity: 98% CAS No.: 504-17-6 Molecular Formula: C4H4N2O2S Formula Weight: 144.15 pH: 1.8 Melting Point: 300° Beilstein Reference: 120663 MDL No.: MFCD00006674

Vzorec: C₄H₄N₂O₂S
Bod varu: 419 °C (1013 hPa)
Bod tání: 235 °C
Hustota: 1,661 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006674
CAS číslo: 504-17-6
EINECS: 207-985-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR