Tisk…

4-Bromobiphenyl (PBB No. 3) ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A12638.09 A12638.18 A12638.30
A12638.09EA 1320 CZK
A12638.09 A12638.18 A12638.30
4-Bromobiphenyl (PBB No. 3) ≥98%
4-Bromobiphenyl (PBB No. 3)

Purity: 98+% CAS No.: 92-66-0 Molecular Formula: C12H9Br Formula Weight: 233.12 Flash Point: 143°(289°F) Melting Point: 88-92° Boiling Point: 310° Beilstein Reference: 1907453 MDL No.: MFCD00000100

Vzorec: C₁₂H₉Br
Bod varu: 310 °C
Bod tání: 88…92 °C
Hustota: 1,615 g/cm³ (20 °C)
Bod vzplanutí: 143 °C (289 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000100
CAS číslo: 92-66-0
EINECS: 202-176-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR