Tisk…

6-Aminobenzothiazole ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A12601.03 A12601.06
A12601.03EA 1790 CZK
A12601.03 A12601.06
6-Aminobenzothiazole ≥98%
6-Aminobenzothiazole

Purity: 98+% CAS No.: 533-30-2 Molecular Formula: C7H6N2S Formula Weight: 150.20 Melting Point: 86-90° Beilstein Reference: 116381 Merck Reference: 14,425 MDL No.: MFCD00015461

Vzorec: C₇H₆N₂S
Bod tání: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00015461
CAS číslo: 533-30-2
EINECS: 208-559-4
Merck Index: 14,425

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR