Tisk…

Triphenylphosphine oxide ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

A12455.14 A12455.22 A12455.36
A12455.14EA 917 CZK
A12455.14 A12455.22 A12455.36
Triphenylphosphine oxide ≥99%
Triphenylphosphine oxide

Purity: 99% CAS No.: 791-28-6 Molecular Formula: C18H15OP Formula Weight: 278.29 Flash Point: 180°(356°F) Melting Point: 153-159° Boiling Point: 360° Beilstein Reference: 745854 MDL No.: MFCD00002080

Vzorec: C₁₈H₁₅OP
Bod varu: 360 °C
Bod tání: 153…159 °C
Hustota: 1,200
Bod vzplanutí: 180 °C (356 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002080
CAS číslo: 791-28-6
EINECS: 212-338-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR