Tisk…

Taurin ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

A12403.0E A12403.22 A12403.36
A12403.0EEA 12460 CZK
A12403.0E A12403.22 A12403.36
Taurin ≥99%
Taurin

Purity: 99% CAS No.: 107-35-7 Molecular Formula: H2NCH2CH2SO3H Formula Weight: 125.15 Physical Form: Crystals or cryst powder Appearance: White Melting Point: 300° Beilstein Reference: 1751215 Merck Reference: 14,9074 MDL No.: MFCD00008197

Vzorec: C₂H₇NO₃S
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00008197
CAS číslo: 107-35-7
EINECS: 203-483-8
Merck Index: 14,9074

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR