Tisk…

Ethyl thiophene-2-carboxylate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A12302.09 A12302.18
A12302.09EA 930 CZK
A12302.09 A12302.18
Ethyl thiophene-2-carboxylate ≥98%
Ethyl thiophene-2-carboxylate

Purity: 98+% CAS No.: 2810-04-0 Molecular Formula: C7H8O2S Formula Weight: 156.20 Flash Point: 88°(190°F) Refractive Index: 1.5260 Boiling Point: 217-218° Beilstein Reference: 112678 MDL No.: MFCD00005436

Vzorec: C₇H₈O₂S
Bod varu: 217…218 °C
Hustota: 1,161
Bod vzplanutí: 88 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005436
CAS číslo: 2810-04-0
EINECS: 220-554-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR