Tisk…

(3-Fluoro-4-hydroxyphenyl)acetic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A12114.06 A12114.03
A12114.06EA 3130 CZK
A12114.06 A12114.03
(3-Fluoro-4-hydroxyphenyl)acetic acid ≥98%
(3-Fluoro-4-hydroxyphenyl)acetic acid

Purity: 98% CAS No.: 458-09-3 Molecular Formula: C8H7FO3 Formula Weight: 170.14 Melting Point: 130-133° MDL No.: MFCD00004348

Vzorec: C₈H₇FO₃
Bod tání: 130…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00004348
CAS číslo: 458-09-3
EINECS: 207-275-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR