Tisk…

N,O-Bis(trimethylsilyl)trifluoroacetamide ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate, Bis(Trimethylsilyl)Trifluoroacetamide, BSTFA

A12033.18 A12033.09 A12033.30
A12033.18EA 7380 CZK
A12033.18 A12033.09 A12033.30
N,O-Bis(trimethylsilyl)trifluoroacetamide ≥98%
N,O-Bis(trimethylsilyl)trifluoroacetamide

Purity: 98+% CAS No.: 25561-30-2 Molecular Formula: C8H18F3NOSi2 Formula Weight: 257.40 Storage Temperature: 2°C to 8°C Flash Point: 23°(73°F) Refractive Index: 1.3840 Melting Point: -10° Boiling Point: 145-147° Beilstein Reference: 3607620 UN No.: 2924 MDL No.: MFCD00008269

Vzorec: C₈H₁₈F₃NOSi₂
Bod varu: 145…147 °C
Bod tání: –10 °C
Hustota: 0,97
Bod vzplanutí: 23 °C (73 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00008269
CAS číslo: 25561-30-2
EINECS: 247-103-9
UN: 2924
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR