Tisk…

Bis(trimethylsilyl)acetylene ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

A11960.09 A11960.18 A11960.30
A11960.09EA 1980 CZK
A11960.09 A11960.18 A11960.30
Bis(trimethylsilyl)acetylene ≥99%
Bis(trimethylsilyl)acetylene

Purity: 99% CAS No.: 14630-40-1 Molecular Formula: C8H18Si2 Formula Weight: 170.40 Flash Point: 2°(35°F) Refractive Index: 1.4270 Melting Point: 20-24° Boiling Point: 134-136° Beilstein Reference: 906870 UN No.: 1993 MDL No.: MFCD00008276

Vzorec: C₈H₁₈Si₂
Bod varu: 134…136 °C
Bod tání: 20…24 °C
Hustota: 0,770
Bod vzplanutí: 2 °C (35 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008276
CAS číslo: 14630-40-1
EINECS: 238-671-9
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR