Tisk…

o-Fenylendiamin 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A11946.30 A11946.0B A11946.0I A11946.14
A11946.30EA 1520 CZK
A11946.30 A11946.0B A11946.0I A11946.14
o-Fenylendiamin 98%
o-Fenylendiamin

Purity: 98% CAS No.: 95-54-5 Molecular Formula: C6H8N2 Formula Weight: 108.14 Flash Point: 136°(277°F) Physical Form: Crystals/flakes Density: 1.270 Sensitivity: Air and Light Sensitive Melting Point: 100-103° Boiling Point: 256-258° Solubility: Soluble in alcohol, chloroform, ether, slightly soluble in water Beilstein Reference: 606074 Merck Reference: 14,7284 UN No.: 1673 MDL No.: MFCD00007721

Vzorec: C₆H₈N₂
Bod varu: 256…258 °C
Bod tání: 100…103 °C
Hustota: 1,270
Bod vzplanutí: 136 °C (277 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007721
CAS číslo: 95-54-5
EINECS: 202-430-6
UN: 1673
ADR: 6.1,III
Merck Index: 14,7284

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR