Tisk…

trans-4-Hydroxy-L(-)-prolin ≥99%

Dodavatel: Thermo Fisher Scientific

A11851.09 A11851.14 A11851.22
A11851.09EA 1590 CZK
A11851.09 A11851.14 A11851.22
trans-4-Hydroxy-L(-)-prolin ≥99%
trans-4-Hydroxy-L(-)-prolin

Purity: 99+% CAS No.: 51-35-4 Molecular Formula: C5H9NO3 Formula Weight: 131.13 Melting Point: 270-273° dec. Beilstein Reference: 471933 Merck Reference: 14,4840 Optical Rotation: -76° (c=4 in water) MDL No.: MFCD00064320

Vzorec: C₅H₉NO₃
Bod varu: 296 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00064320
CAS číslo: 51-35-4
EINECS: 200-091-9
Merck Index: 14,4840

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR