Tisk…

N-Etyldiizopropylamin 99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Hünig’s base, N,N-Diisopropylethylamine, N.N’-Diisopropyl ethylamine, DIPEA

A11801.AEEA 1500 CZK
A11801.AE A11801.AC A11801.AP A11801.AU
N-Etyldiizopropylamin 99%
N-Etyldiizopropylamin

Purity: 99% CAS No.: 7087-68-5 Molecular Formula: C8H19N Formula Weight: 129.25 Flash Point: 8°(46°F) Physical Form: Liquid Density: 0.747 Refractive Index: 1.4135 Appearance: Colourless to pale yellow Melting Point: -46° Boiling Point: 126-127° Beilstein Reference: 605301 UN No.: 3488 MDL No.: MFCD00008868

Vzorec: C₈H₁₉N
MW: 129.25
Bod varu: 126…127 °C
Bod tání: –46 °C
Hustota: 0.76
Bod vzplanutí: 8 °C (46 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008868
CAS číslo: 7087-68-5
EINECS: 230-392-0
UN: 3384
ADR: 6.1 I

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR