Tisk…

Thiophene-2-methylamine 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A11700.06 A11700.14 A11700.22
A11700.06EA 1570 CZK
A11700.06 A11700.14 A11700.22
Thiophene-2-methylamine 97%
Thiophene-2-methylamine

Purity: 97% CAS No.: 27757-85-3 Molecular Formula: C5H7NS Formula Weight: 113.19 Flash Point: 73°(163°F) Refractive Index: 1.5670 Boiling Point: 85-88°/14mm Beilstein Reference: 106296 UN No.: 2735 MDL No.: MFCD00005460

Vzorec: C₅H₇NS
Bod varu: 85…88 °C (14 mmHg)
Hustota: 1,100
Bod vzplanutí: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005460
CAS číslo: 27757-85-3
EINECS: 248-639-6
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR