Tisk…

2-Chloro-3-hydroxypyridine ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A11376.14 A11376.22
A11376.14EA 4360 CZK
A11376.14 A11376.22
2-Chloro-3-hydroxypyridine ≥98%
2-Chloro-3-hydroxypyridine

Purity: 98% CAS No.: 6636-78-8 Molecular Formula: C5H4ClNO Formula Weight: 129.55 Melting Point: 168-172° Beilstein Reference: 2216

Vzorec: C₅H₄ClNO
Bod varu: 212 °C (1013 hPa)
Bod tání: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00006235
CAS číslo: 6636-78-8
EINECS: 229-635-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR