Tisk…

2,6-Dichlorobenzaldehyde oxime ≥97%

Dodavatel: Thermo Fisher Scientific

Synonyms: 2,6-Dichlorobenzaldoxime

A11351.06 A11351.14 A11351.22
A11351.06EA 927 CZK
A11351.06 A11351.14 A11351.22
2,6-Dichlorobenzaldehyde oxime ≥97%
2,6-Dichlorobenzaldehyde oxime

Purity: 97% CAS No.: 25185-95-9 Molecular Formula: C7H5Cl2NO Formula Weight: 190.03 Melting Point: 147-151° Beilstein Reference: 2046662

Vzorec: C₇H₅Cl₂NO
MW: 190.03
Bod tání: 147…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00013938
CAS číslo: 25185-95-9
EINECS: 246-720-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR