Tisk…

Dihydrogenfosforečnan sodný dihydrát ≥99%

Dodavatel: Thermo Fisher Scientific

Synonyms: Phosphoric acid (sodium hydrate), Sodium phosphate monobasic dihydrate, Sodium dihydrogen orthophosphate dihydrate

A11316.0E A11316.36 A11316.22
A11316.0EEA 3540 CZK
A11316.0E A11316.36 A11316.22
Dihydrogenfosforečnan sodný dihydrát ≥99%
Dihydrogenfosforečnan sodný dihydrát

Purity: 99% CAS No.: 13472-35-0 Molecular Formula: H2NaO4P.2H2O Formula Weight: 156.01 (119.98anhy) Merck Reference: 14,8660 MDL No.: MFCD00149209

Vzorec: NaH₂PO₄·2H₂O
MW: 156.01
Bod varu: ∼650 °C (1013 hPa)
Bod tání: 60
Hustota: 1.910
Storage Temperature: Ambient
MDL Number: MFCD00149209
CAS číslo: 13472-35-0
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR