Tisk…

(3-Aminopropyl)trimethoxysilane 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A11284.0E A11284.22 A11284.36
A11284.0EEA 26600 CZK
A11284.0E A11284.22 A11284.36
(3-Aminopropyl)trimethoxysilane 97%
(3-Aminopropyl)trimethoxysilane

Purity: 97% CAS No.: 13822-56-5 Molecular Formula: C6H17NO3Si Formula Weight: 179.29 Flash Point: 90°(194°F) Physical Form: Liquid Density: 1.02 Refractive Index: 1.4240 Sensitivity: Air and Moisture Sensitive Appearance: Colourless Odour: Amine-like Boiling Point: 194° Beilstein Reference: 2038118 UN No.: 2735 MDL No.: MFCD00008206

Vzorec: H₂NCH₂CH₂CH₂Si(OCH₃)₃
Bod varu: 194 °C
Hustota: 1,02
Bod vzplanutí: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008206
CAS číslo: 13822-56-5
EINECS: 237-511-5
UN: 2735
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR