Tisk…

D-(-)-Tartaric acid ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: unnatural tartaric acid, (2S,3S)-2,3-Dihydroxysuccinic acid, (S,S)-(-)-Tartaric acid, (S,S)-(-)-2,3-Dihydroxysuccinic acid

A11264.14 A11264.22 A11264.36
A11264.14EA 1540 CZK
A11264.14 A11264.22 A11264.36
D-(-)-Tartaric acid ≥99%
D-(-)-Tartaric acid

Purity: 99% CAS No.: 147-71-7 Molecular Formula: C4H6O6 Formula Weight: 150.09 Flash Point: 210°(410°F) Odour: Odourless pH: 1.6 Melting Point: 166-170° Beilstein Reference: 1725145 Merck Reference: 14,9068 Optical Rotation: -13° (c=10 in water) MDL No.: MFCD00004238

Vzorec: C₄H₆O₆
Bod varu: 356 °C (1013 hPa)
Bod tání: 166…170 °C
Hustota: 1,76 g/cm³ (20 °C)
Bod vzplanutí: 210 °C (410 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004238
CAS číslo: 147-71-7
EINECS: 205-695-6
Merck Index: 14,9068

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR