Tisk…

2-Methyl-1,3-cyclohexanedione ≥98%

Dodavatel: Thermo Fisher Scientific

A11218.06 A11218.14 A11218.22
A11218.06EA 1160 CZK
A11218.06 A11218.14 A11218.22
2-Methyl-1,3-cyclohexanedione ≥98%
2-Methyl-1,3-cyclohexanedione

Purity: 98+% CAS No.: 1193-55-1 Molecular Formula: C7H10O2 Formula Weight: 126.16 Storage Temperature: 2°C to 8°C Melting Point: 204-208° Beilstein Reference: 1281598 MDL No.: MFCD00001587

Vzorec: C₇H₁₀O₂
Bod varu: 100…102 °C (8 mmHg)
Bod tání: 204…208 °C
Hustota: 1,313 g/cm³ (–150.15 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001587
CAS číslo: 1193-55-1
EINECS: 214-773-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR