Tisk…

2'-Deoxyadenosine monohydrate ≥99%

Dodavatel: Thermo Fisher Scientific

Synonyms: (2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol hydrate

A11166.03 A11166.06 A11166.14
A11166.03EA 1500 CZK
A11166.03 A11166.06 A11166.14
2'-Deoxyadenosine monohydrate ≥99%
2'-Deoxyadenosine monohydrate

Purity: 99% CAS No.: 16373-93-6 Molecular Formula: C10H13N5O3.H2O Formula Weight: 269.26 (251.24anhy) Storage Temperature: 2°C to 8°C Melting Point: 186-189° Beilstein Reference: 5191174 Optical Rotation: -25° (c=0.5 water) MDL No.: MFCD00149364

Vzorec: C₁₀H₁₃N₅O₃·1H₂O
Bod tání: 186…189 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149364
CAS číslo: 16373-93-6
EINECS: 213-488-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR