Tisk…

2-Nitroanilin ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: o-Nitroaniline

A11055.0B A11055.0I A11055.30
A11055.0BEA 2750 CZK
A11055.0B A11055.0I A11055.30
2-Nitroanilin ≥98%
2-Nitroanilin

Purity: 98% CAS No.: 88-74-4 Molecular Formula: C6H6N2O2 Formula Weight: 138.13 Flash Point: 168°(334°F) Melting Point: 70-74° Boiling Point: 284-286° Beilstein Reference: 509275 Merck Reference: 14,6583 UN No.: 1661 MDL No.: MFCD00007687

Vzorec: C₆H₆N₂O₂
Bod varu: 284…286 °C
Bod tání: 70…74 °C
Hustota: 1,255
Bod vzplanutí: 168 °C (334 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007687
CAS číslo: 88-74-4
EINECS: 201-855-4
UN: 1661
ADR: 6.1,II
Merck Index: 14,6583

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR