Tisk…

(±)-Kafr 96%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: (±)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, Camphor, 1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, DL-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (±)-1,7,7-Trimethyl-2-norbornanone

A10936.0C A10936.0E A10936.22 A10936.36
A10936.0CEA 13460 CZK
A10936.0C A10936.0E A10936.22 A10936.36
(±)-Kafr 96%
(±)-Kafr

Purity: 96% CAS No.: 76-22-2 Molecular Formula: C10H16O Formula Weight: 152.24 Flash Point: 64°(147°F) Physical Form: Crystalline powder or lumps Density: 0.992 Appearance: White Odour: Camphor-like Melting Point: 172-176° Boiling Point: 204° Solubility: Soluble in alcohols, ether, chloroform, benzene Beilstein Reference: 1907611 Merck Reference: 14,1732 UN No.: 2717 MDL No.: MFCD00074738

Vzorec: C₁₀H₁₆O
MW: 152.24
Bod varu: 204 °C
Bod tání: 172…176 °C
Hustota: 0.992
Bod vzplanutí: 64 °C (147 °F)
Storage Temperature: Ambient
MDL Number: MFCD00074738
CAS číslo: 76-22-2
EINECS: 200-945-0
UN: 2717
ADR: 4.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR