Tisk…

Ethyl (S)-(-)-laktát ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A10900.36 A10900.0C A10900.0E
A10900.36EA 1170 CZK
A10900.36 A10900.0C A10900.0E
Ethyl (S)-(-)-laktát ≥99%
Ethyl (S)-(-)-laktát

Purity: 99% CAS No.: 687-47-8 Molecular Formula: C5H10O3 Formula Weight: 118.13 Flash Point: 46°(114°F) Physical Form: Liquid Density: 1.036 Refractive Index: 1.4130 Appearance: Colourless Melting Point: -25° Boiling Point: 152-155° Beilstein Reference: 1720839 Merck Reference: 14,3817 Optical Rotation: -11° (neat) UN No.: 1192 MDL No.: MFCD00065359

Vzorec: C₅H₁₀O₃
Bod varu: 152…155 °C
Bod tání: –25 °C
Hustota: 1,036
Bod vzplanutí: 46 °C (114 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004518
CAS číslo: 687-47-8
EINECS: 211-694-1
UN: 1192
ADR: 3,III
Merck Index: 14,3817

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR