Tisk…

Tetrabutylphosphonium bromide ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A10868.14 A10868.22 A10868.36
A10868.14EA 924 CZK
A10868.14 A10868.22 A10868.36
Tetrabutylphosphonium bromide ≥99%
Tetrabutylphosphonium bromide

Purity: 99% CAS No.: 3115-68-2 Molecular Formula: C16H36BrP Formula Weight: 339.35 Flash Point: 290°(554°F) Melting Point: 99-103° Beilstein Reference: 4160474 UN No.: 3464 MDL No.: MFCD00011853

Vzorec: C₁₆H₃₆BrP
Bod tání: 99…103 °C
Bod vzplanutí: 290 °C (554 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011853
CAS číslo: 3115-68-2
EINECS: 221-487-8
UN: 3464
ADR: 6.1,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR