Tisk…

4-Bromo-4'-hydroxybiphenyl ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 4'-Bromo-[1,1'-biphenyl]-4-ol

A10819.06 A10819.14 A10819.22
A10819.06EA 2160 CZK
A10819.06 A10819.14 A10819.22
4-Bromo-4'-hydroxybiphenyl ≥98%
4-Bromo-4'-hydroxybiphenyl

Purity: 98% CAS No.: 29558-77-8 Molecular Formula: C12H9BrO Formula Weight: 249.11 Melting Point: 163-168° MDL No.: MFCD00059076

Vzorec: C₁₂H₉BrO
MW: 249.11 g/mol
Bod tání: 163…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00059076
CAS číslo: 29558-77-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR