Tisk…

Ruthenium(IV) oxide, anhydrous 99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Ruthenium dioxide, Ruthenium tetroxide

A10816.14 A10816.03 A10816.06
A10816.14EA 70690 CZK
A10816.14 A10816.03 A10816.06
Ruthenium(IV) oxide, anhydrous 99.9% (metals basis)
Ruthenium(IV) oxide

Purity: 99.9% CAS No.: 12036-10-1 Molecular Formula: RuO2 Formula Weight: 133.07 Odour: Odourless Melting Point: 1200° subl. UN No.: 1479 MDL No.: MFCD00011210

Vzorec: O₂Ru
Bod tání: 1200 °C (subl.)
Hustota: 6,97
Storage Temperature: Ambient
MDL Number: MFCD00011210
CAS číslo: 12036-10-1
EINECS: 234-840-6
UN: 1479
ADR: 5.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR