Tisk…

Acetone oxime ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Propan-2-one oxime, Acetoxime, Acetone oxime, 2-Propanone oxime

A10802.0B A10802.18 A10802.30
A10802.0BEA 7900 CZK
A10802.0B A10802.18 A10802.30
Acetone oxime ≥98%
Acetone oxime

Purity: 98% CAS No.: 127-06-0 Molecular Formula: C3H7NO Formula Weight: 73.10 Flash Point: 60°(140°F) Melting Point: 60-63° Boiling Point: 133-135° Beilstein Reference: 1560146 Merck Reference: 14,75 UN No.: 1325 MDL No.: MFCD00002118

Vzorec: C₃H₇NO
MW: 73.09
Bod varu: 133…135 °C
Bod tání: 60…63 °C
Hustota: 1.045
Bod vzplanutí: 60 °C (140 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002118
CAS číslo: 127-06-0
EINECS: 204-820-1
UN: 1325
ADR: 4.1 III
Merck Index: 13,00077

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR