Tisk…

Tributyltin chloride 96%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A10746.0B A10746.18 A10746.30
A10746.0BEA 8410 CZK
A10746.0B A10746.18 A10746.30
Tributyltin chloride 96%
Tributyltin chloride

Purity: 96% CAS No.: 1461-22-9 Molecular Formula: C12H27ClSn Formula Weight: 325.51 Flash Point: 120°(248°F) Refractive Index: 1.4910 Melting Point: -19° Boiling Point: 171-173°/25mm Beilstein Reference: 3535715 UN No.: 2788 MDL No.: MFCD00000521

Vzorec: C₁₂H₂₇ClSn
Bod varu: 171…173 °C (25 mmHg)
Bod tání: –19 °C
Hustota: 1,200
Bod vzplanutí: 120 °C (248 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000521
CAS číslo: 1461-22-9
EINECS: 215-958-7
UN: 2788
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR