Tisk…

2,5-Difluorobenzoyl chloride ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A10651.06 A10651.14
A10651.06EA 1600 CZK
A10651.06 A10651.14
2,5-Difluorobenzoyl chloride ≥98%
2,5-Difluorobenzoyl chloride

Purity: 98% CAS No.: 35730-09-7 Molecular Formula: C7H3ClF2O Formula Weight: 176.55 Flash Point: 93°(199°F) Refractive Index: 1.5150 Boiling Point: 81-83°/14mm Beilstein Reference: 2046666 UN No.: 3265 MDL No.: MFCD00009929

Vzorec: C₇H₃ClF₂O
MW: 176.55
Bod varu: 81…83 °C (14 mmHg)
Hustota: 1.42
Bod vzplanutí: 93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009929
CAS číslo: 35730-09-7
EINECS: 252-701-8
UN: 3265
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR