Tisk…

3-Chloropropanesulphonyl chloride ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A10631.09 A10631.18 A10631.30
A10631.09EA 2820 CZK
A10631.09 A10631.18 A10631.30
3-Chloropropanesulphonyl chloride ≥98%
3-Chloropropanesulphonyl chloride

Purity: 98+% CAS No.: 1633-82-5 Molecular Formula: C3H6Cl2O2S Formula Weight: 177.05 Flash Point: 110°(230°F) Refractive Index: 1.4900 Boiling Point: 70°/0.5mm Beilstein Reference: 1754119 UN No.: UN3265 MDL No.: MFCD00007463

Vzorec: C₃H₆Cl₂O₂S
Bod varu: 70 °C (0,5 mmHg)
Hustota: 1,456
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007463
CAS číslo: 1633-82-5
EINECS: 216-646-3
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR