Tisk…

Dimethyl-2-aminoterephthalate 99%

Dodavatel: Thermo Fisher Scientific
Varování

A10567.0B A10567.18 A10567.30
A10567.0BEA 21070 CZK
A10567.0B A10567.18 A10567.30
Dimethyl-2-aminoterephthalate 99%
Dimethyl-2-aminoterephthalate

Purity: 99% CAS No.: 5372-81-6 Molecular Formula: C10H11NO4 Formula Weight: 209.20 Melting Point: 130-134° Beilstein Reference: 2808505

Vzorec: C₁₀H₁₁NO₄
Bod tání: 130…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00008427
CAS číslo: 5372-81-6
EINECS: 226-364-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR