Tisk…

1,4-Dibromo-2-nitrobenzene ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2,5-Dibromonitrobenzene

A10472.14 A10472.22
A10472.14EA 1070 CZK
A10472.14 A10472.22
1,4-Dibromo-2-nitrobenzene ≥99%
1,4-Dibromo-2-nitrobenzene

Purity: 99% CAS No.: 3460-18-2 Molecular Formula: C6H3Br2NO2 Formula Weight: 280.91 Melting Point: 82-85° Beilstein Reference: 1950425

Vzorec: C₆H₃Br₂NO₂
Bod tání: 82…85 °C
Hustota: 2,374
Storage Temperature: Ambient
MDL Number: MFCD00007046
CAS číslo: 3460-18-2
EINECS: 222-404-8
UN: 3077
ADR: 9,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR