Tisk…

4-Methylumbelliferone ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

A10337.0B A10337.18 A10337.30
A10337.0BEA 11470 CZK
A10337.0B A10337.18 A10337.30
4-Methylumbelliferone ≥97%
4-Methylumbelliferone

Purity: 97% CAS No.: 90-33-5 Molecular Formula: C10H8O3 Formula Weight: 176.17 Melting Point: 186-191° Solubility: Soluble in alcohol. Insoluble in cold water Beilstein Reference: 142217 Merck Reference: 14,4854 MDL No.: MFCD00006866

Vzorec: C₁₀H₈O₃
Bod tání: 186…191 °C
Storage Temperature: Ambient
MDL Number: MFCD00006866
CAS číslo: 90-33-5
EINECS: 201-986-7
Merck Index: 14,4854

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR